Imprimer

Anti-ATP7B Rabbit Polyclonal Antibody (DyLight® 650)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-361C
NOVUNB100-361CEA 656 EUR
NOVUNB100-361C
Anti-ATP7B Rabbit Polyclonal Antibody (DyLight® 650)
Anticorps
Type: Primary
Antigen: ATP7B (ATPase, Cu++ transporting, beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR