Imprimer

Anti-RAD21 Mouse Monoclonal Antibody (DyLight® 488) [clone: CM110-2C10]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-386GEA 656 EUR
NOVUNB100-386G
Anti-RAD21 Mouse Monoclonal Antibody (DyLight® 488) [clone: CM110-2C10]
Anticorps
Type: Primary
Antigen: RAD21 (RAD21 homolog (S. pombe))
Clonality: monoclonal
Clone: CM110-2C10
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR