Imprimer

Anti-Separase Mouse Monoclonal Antibody (DyLight® 488) [clone: XJ11-1B12]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-439GEA 613 EUR
NOVUNB100-439G
Anti-Separase Mouse Monoclonal Antibody (DyLight® 488) [clone: XJ11-1B12]
Anticorps
Type: Primary
Antigen: Separase
Clonality: monoclonal
Clone: XJ11-1B12
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR