Imprimer

Anti-FLT1 Rabbit Polyclonal Antibody (DyLight® 650) [clone: 4C11]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-527CEA 663 EUR
NOVUNB100-527C
Anti-FLT1 Rabbit Polyclonal Antibody (DyLight® 650) [clone: 4C11]
Anticorps
Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: polyclonal
Clone: 4C11
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR