Imprimer

Anti-CASP8 Mouse Monoclonal Antibody (Biotin) [clone: 90A992]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-55786
NOVUNB100-55786EA 698 EUR
NOVUNB100-55786
Anti-CASP8 Mouse Monoclonal Antibody (Biotin) [clone: 90A992]
Anticorps
Type: Primary
Antigen: CASP8 (caspase 8, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: 90A992
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR