Imprimer

Anti-TNFRSF11A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9A725]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-56029EA 669 EUR
NOVUNB100-56029
Anti-TNFRSF11A Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 9A725]
Anticorps
Type: Primary
Antigen: TNFRSF11A (tumor necrosis factor receptor superfamily, member 11a, NFKB activator)
Clonality: monoclonal
Clone: 9A725
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR