Imprimer

Anti-TLR4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HTA125]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-56062EA 652 EUR
NOVUNB100-56062
Anti-TLR4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HTA125]
Anticorps
Type: Primary
Antigen: TLR4 (toll-like receptor 4)
Clonality: monoclonal
Clone: HTA125
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR