Imprimer

Anti-TNFRSF11A Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-56396
NOVUNB100-56396EA 546 EUR
NOVUNB100-56396
Anti-TNFRSF11A Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TNFRSF11A (tumor necrosis factor receptor superfamily, member 11a, NFKB activator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR