Imprimer

Anti-TNFSF10 Mouse Monoclonal Antibody [clone: 55B709.3]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-56518EA 586 EUR
NOVUNB100-56518
Anti-TNFSF10 Mouse Monoclonal Antibody [clone: 55B709.3]
Anticorps
Type: Primary
Antigen: TNFSF10 (tumor necrosis factor (ligand) superfamily, member 10)
Clonality: monoclonal
Clone: 55B709.3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR