Imprimer

Anti-RAD18 Mouse Monoclonal Antibody [clone: 79B1048.1]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-56523
NOVUNB100-56523EA 586 EUR
NOVUNB100-56523
Anti-RAD18 Mouse Monoclonal Antibody [clone: 79B1048.1]
Anticorps
Type: Primary
Antigen: RAD18 (RAD18 homolog (S. cerevisiae))
Clonality: monoclonal
Clone: 79B1048.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR