Imprimer

Anti-STAM Mouse Monoclonal Antibody [clone: 29C678]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-56533
NOVUNB100-56533EA 586 EUR
NOVUNB100-56533
Anti-STAM Mouse Monoclonal Antibody [clone: 29C678]
Anticorps
Type: Primary
Antigen: STAM (signal transducing adaptor molecule (SH3 domain and ITAM motif) 1)
Clonality: monoclonal
Clone: 29C678
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR