Imprimer

Anti-PMS2 Mouse Monoclonal Antibody [clone: 163C1251]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-56554
NOVUNB100-56554EA 425 EUR
NOVUNB100-56554
Anti-PMS2 Mouse Monoclonal Antibody [clone: 163C1251]
Anticorps
Type: Primary
Antigen: PMS2 (PMS2 postmeiotic segregation increased 2 (S. cerevisiae))
Clonality: monoclonal
Clone: 163C1251
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR