Imprimer

Anti-PLXDC1 Mouse Monoclonal Antibody [clone: 197C193 (IM193)]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-56557EA 602 EUR
NOVUNB100-56557
Anti-PLXDC1 Mouse Monoclonal Antibody [clone: 197C193 (IM193)]
Anticorps
Type: Primary
Antigen: PLXDC1 (plexin domain containing 1)
Clonality: monoclonal
Clone: 197C193 (IM193)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR