Imprimer

Anti-HTRA2 Mouse Monoclonal Antibody [clone: 196C429]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-56558EA 548 EUR
NOVUNB100-56558
Anti-HTRA2 Mouse Monoclonal Antibody [clone: 196C429]
Anticorps
Type: Primary
Antigen: HTRA2 (HtrA serine peptidase 2)
Clonality: monoclonal
Clone: 196C429
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR