Imprimer

Anti-DNA Ligase I Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-56635
NOVUNB100-56635EA 425 EUR
NOVUNB100-56635
Anti-DNA Ligase I Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: DNA Ligase I
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR