Imprimer
Anti-ASB11 Rabbit Polyclonal Antibody
Fournisseur: Novus Biologicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NOVUNB100-56640EA 398 EUR
NOVUNB100-56640
Anti-ASB11 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: ASB11 (ankyrin repeat and SOCS box containing 11, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human