Imprimer

Anti-GABRG3 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-56662EA 406 EUR
NOVUNB100-56662
Anti-GABRG3 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: GABRG3 (gamma-aminobutyric acid (GABA) A receptor, gamma 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR