Imprimer

Anti-AKT2 Mouse Monoclonal Antibody [clone: 95C657.1.2]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-56676
NOVUNB100-56676EA 425 EUR
NOVUNB100-56676
Anti-AKT2 Mouse Monoclonal Antibody [clone: 95C657.1.2]
Anticorps
Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: monoclonal
Clone: 95C657.1.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR