Imprimer

Anti-CASP3 Mouse monoclonal antibody unconjugated [clone: 31A1067]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-56708EA 586 EUR
NOVUNB100-56708
Anti-CASP3 Mouse monoclonal antibody unconjugated [clone: 31A1067]
Anticorps
Type: Primary
Antigen: CASP3 (caspase 3, apoptosis-related cysteine peptidase)
Clonality: monoclonal
Clone: 31A1067
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR