Imprimer

Anti-TRAF3IP2 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-56740
NOVUNB100-56740EA 546 EUR
NOVUNB100-56740
Anti-TRAF3IP2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TRAF3IP2 (TRAF3 interacting protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR