Imprimer

Anti-KMO Mouse Monoclonal Antibody (Biotin) [clone: P3UI]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-597BEA 656 EUR
NOVUNB100-597B
Anti-KMO Mouse Monoclonal Antibody (Biotin) [clone: P3UI]
Anticorps
Type: Primary
Antigen: KMO (kynurenine 3-monooxygenase (kynurenine 3-hydroxylase))
Clonality: monoclonal
Clone: P3UI
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR