Imprimer

Anti-BRCA1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RAY]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-598APCEA 613 EUR
NOVUNB100-598APC
Anti-BRCA1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RAY]
Anticorps
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: RAY
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR