Imprimer

Anti-BRCA1 Mouse Monoclonal Antibody (DyLight® 650) [clone: MU]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-600CEA 709 EUR
NOVUNB100-600C
Anti-BRCA1 Mouse Monoclonal Antibody (DyLight® 650) [clone: MU]
Anticorps
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: MU
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR