Imprimer

Anti-CAV1 Mouse Monoclonal Antibody (DyLight® 350) [clone: 7C8]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-615UVEA 737 EUR
NOVUNB100-615UV
Anti-CAV1 Mouse Monoclonal Antibody (DyLight® 350) [clone: 7C8]
Anticorps
Type: Primary
Antigen: CAV1 (caveolin 1, caveolae protein, 22kDa)
Clonality: monoclonal
Clone: 7C8
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR