Imprimer

Anti-6X His tag Mouse Monoclonal Antibody (DyLight® 755) [clone: AD1.1.10]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-64768IREA 709 EUR
NOVUNB100-64768IR
Anti-6X His tag Mouse Monoclonal Antibody (DyLight® 755) [clone: AD1.1.10]
Anticorps
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: DyLight® 755
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Pan
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR