Imprimer

Anti-VEGFA Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: VG76e]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-648H
NOVUNB100-648HEA 826 EUR
NOVUNB100-648H
Anti-VEGFA Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: VG76e]
Anticorps
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: VG76e
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR