Imprimer

Anti-VEGFA Mouse Monoclonal Antibody (DyLight® 550) [clone: VG1]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-664REA 739 EUR
NOVUNB100-664R
Anti-VEGFA Mouse Monoclonal Antibody (DyLight® 550) [clone: VG1]
Anticorps
Type: Primary
Antigen: VEGFA (vascular endothelial growth factor A)
Clonality: monoclonal
Clone: VG1
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR