Imprimer

Anti-TUBA Mouse Monoclonal Antibody (DyLight® 350) [clone: DM1A]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-690UVEA 596 EUR
NOVUNB100-690UV
Anti-TUBA Mouse Monoclonal Antibody (DyLight® 350) [clone: DM1A]
Anticorps
Type: Primary
Antigen: TUBA (tubulin, alpha)
Clonality: monoclonal
Clone: DM1A
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR