Imprimer

Anti-LYVE1 Rabbit Polyclonal Antibody (DyLight® 650)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-726C
NOVUNB100-726CEA 485 EUR
NOVUNB100-726C
Anti-LYVE1 Rabbit Polyclonal Antibody (DyLight® 650)
Anticorps
Type: Primary
Antigen: LYVE1 (lymphatic vessel endothelial hyaluronan receptor 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR