Imprimer

Anti-Aggrecan Neoepitope Rabbit Polyclonal Antibody (DyLight® 488)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB100-74350GEA 663 EUR
NOVUNB100-74350G
Anti-Aggrecan Neoepitope Rabbit Polyclonal Antibody (DyLight® 488)
Anticorps
Type: Primary
Antigen: Aggrecan Neoepitope (aggrecan Neoepitope)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR