Imprimer

Anti-TFRC Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB100-92243
NOVUNB100-92243EA 620 EUR
NOVUNB100-92243
Anti-TFRC Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TFRC (transferrin receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR