Imprimer

Anti-SLC2A1 Rabbit Polyclonal Antibody (DyLight® 550)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-39113REA 739 EUR
NOVUNB110-39113R
Anti-SLC2A1 Rabbit Polyclonal Antibody (DyLight® 550)
Anticorps
Type: Primary
Antigen: SLC2A1 (solute carrier family 2 (facilitated glucose transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR