Imprimer

Anti-TRPA1 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-40763 NB110-40763SS
NOVUNB110-40763EA 702 EUR
NOVUNB110-40763 NOVUNB110-40763SS
Anti-TRPA1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TRPA1 (transient receptor potential cation channel, subfamily A, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR