Imprimer

Anti-KCNK2 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-41535 NB110-41535SS
NOVUNB110-41535EA 580 EUR
NOVUNB110-41535 NOVUNB110-41535SS
Anti-KCNK2 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: KCNK2 (potassium channel, subfamily K, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR