Imprimer

Anti-CITED4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HT13-2D6.3]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-41572PEEA 613 EUR
NOVUNB110-41572PE
Anti-CITED4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HT13-2D6.3]
Anticorps
Type: Primary
Antigen: CITED4 (Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4)
Clonality: monoclonal
Clone: HT13-2D6.3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR