Imprimer

Anti-DNM1L Rabbit Polyclonal Antibody (DyLight® 405)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-55288VEA 652 EUR
NOVUNB110-55288V
Anti-DNM1L Rabbit Polyclonal Antibody (DyLight® 405)
Anticorps
Type: Primary
Antigen: DNM1L (dynamin 1-like)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR