Imprimer

Anti-APOA5 Mouse Monoclonal Antibody (DyLight® 650) [clone: 1G5G9]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-55454CEA 613 EUR
NOVUNB110-55454C
Anti-APOA5 Mouse Monoclonal Antibody (DyLight® 650) [clone: 1G5G9]
Anticorps
Type: Primary
Antigen: APOA5 (apolipoprotein A-V)
Clonality: monoclonal
Clone: 1G5G9
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR