Imprimer

Anti-TERF2 Rabbit Polyclonal Antibody (DyLight® 650)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-57130CEA 705 EUR
NOVUNB110-57130C
Anti-TERF2 Rabbit Polyclonal Antibody (DyLight® 650)
Anticorps
Type: Primary
Antigen: TERF2 (telomeric repeat binding factor 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR