Imprimer

Anti-TMED10 Rabbit Polyclonal Antibody (DyLight® 550)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-57586R
NOVUNB110-57586REA 656 EUR
NOVUNB110-57586R
Anti-TMED10 Rabbit Polyclonal Antibody (DyLight® 550)
Anticorps
Type: Primary
Antigen: TMED10 (transmembrane emp24-like trafficking protein 10 (yeast))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR