Imprimer

Anti-ALOX5 Rabbit Polyclonal Antibody (DyLight® 488)

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-58749GEA 555 EUR
NOVUNB110-58749G
Anti-ALOX5 Rabbit Polyclonal Antibody (DyLight® 488)
Anticorps
Type: Primary
Antigen: ALOX5 (arachidonate 5-lipoxygenase)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR