Imprimer

Anti-PMP22 Mouse Monoclonal Antibody (DyLight® 550) [clone: Hu1]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-59086REA 613 EUR
NOVUNB110-59086R
Anti-PMP22 Mouse Monoclonal Antibody (DyLight® 550) [clone: Hu1]
Anticorps
Type: Primary
Antigen: PMP22 (peripheral myelin protein 22)
Clonality: monoclonal
Clone: Hu1
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR