Imprimer

Anti-KIAA1524 Mouse Monoclonal Antibody (DyLight® 405) [clone: 2G10]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-59722VEA 663 EUR
NOVUNB110-59722V
Anti-KIAA1524 Mouse Monoclonal Antibody (DyLight® 405) [clone: 2G10]
Anticorps
Type: Primary
Antigen: KIAA1524 (KIAA1524)
Clonality: monoclonal
Clone: 2G10
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR