Imprimer

Anti-CD36 Mouse Monoclonal Antibody (DyLight® 405) [clone: D-2712]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-59724VEA 671 EUR
NOVUNB110-59724V
Anti-CD36 Mouse Monoclonal Antibody (DyLight® 405) [clone: D-2712]
Anticorps
Type: Primary
Antigen: CD36 (CD36 molecule (thrombospondin receptor))
Clonality: monoclonal
Clone: D-2712
Conjugation: DyLight® 405
Epitope:
Host: Mouse
Isotype: IgA Kappa
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR