Imprimer

Anti-DEFA5 Mouse Monoclonal Antibody (Biotin) [clone: 8C8]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-60002B
NOVUNB110-60002BEA 722 EUR
NOVUNB110-60002B
Anti-DEFA5 Mouse Monoclonal Antibody (Biotin) [clone: 8C8]
Anticorps
The Defensin alpha 5 Antibody (8C8) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to Defensin alpha 5. This antibody reacts with human. The Defensin alpha 5 Antibody (8C8) [Biotin] has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5-10 ?g/ml

Type: Primary
Antigen: DEFA5
Clonality: Monoclonal
Clone: 8C8
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR