Imprimer

Anti-DEFA5 Mouse Monoclonal Antibody (DyLight® 680) [clone: 8C8]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-60002FREA 663 EUR
NOVUNB110-60002FR
Anti-DEFA5 Mouse Monoclonal Antibody (DyLight® 680) [clone: 8C8]
Anticorps
Type: Primary
Antigen: DEFA5 (defensin, alpha 5, Paneth cell-specific)
Clonality: monoclonal
Clone: 8C8
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR