Imprimer

Anti-DRD1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: SG2-D1a]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-60017FEA 663 EUR
NOVUNB110-60017F
Anti-DRD1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: SG2-D1a]
Anticorps
Type: Primary
Antigen: DRD1 (dopamine receptor D1)
Clonality: monoclonal
Clone: SG2-D1a
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR