Imprimer

Anti-DRD5 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: SG4-D1b]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NOVUNB110-60019HEA 663 EUR
NOVUNB110-60019H
Anti-DRD5 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: SG4-D1b]
Anticorps
Type: Primary
Antigen: DRD5 (dopamine receptor D5)
Clonality: monoclonal
Clone: SG4-D1b
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR