Imprimer

Anti-BAP1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 1G8]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-60521F
NOVUNB110-60521FEA 709 EUR
NOVUNB110-60521F
Anti-BAP1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 1G8]
Anticorps
Type: Primary
Antigen: BAP1 (BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase))
Clonality: monoclonal
Clone: 1G8
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR