Imprimer

Anti-APOE Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: WUE-4]

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-60531H
NOVUNB110-60531HEA 826 EUR
NOVUNB110-60531H
Anti-APOE Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: WUE-4]
Anticorps
Type: Primary
Antigen: APOE (apolipoprotein E)
Clonality: monoclonal
Clone: WUE-4
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR