Imprimer

Anti-STIM1 Rabbit Polyclonal Antibody

Fournisseur: Novus Biologicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

NB110-60547 NB110-60547SS
NOVUNB110-60547EA 574 EUR
NOVUNB110-60547 NOVUNB110-60547SS
Anti-STIM1 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: STIM1 (stromal interaction molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR